Assay Details
Gene Symbol: WDR53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716531.3 | XP_002716577.1 | 2 - 3 | 528 | 104 | |
XM_017347015.1 | XP_017202504.1 | 2 - 3 | 861 | 104 | ||
XM_017347016.1 | XP_017202505.1 | 3 - 4 | 781 | 104 | ||
XM_017347017.1 | XP_017202506.1 | 2 - 3 | 605 | 104 | ||
XM_017347018.1 | XP_017202507.1 | 2 - 3 | 522 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
