Assay Details
| Gene Symbol: C16H1orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717335.2 | XP_002717381.1 | 3 - 4 | 584 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16H1orf131| Gene Symbol: C16H1orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717335.2 | XP_002717381.1 | 3 - 4 | 584 | 70 | |
Set Membership: |
|