Assay Details
Gene Symbol: C16H1orf131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717335.2 | XP_002717381.1 | 6 - 7 | 814 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C16H1orf131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717335.2 | XP_002717381.1 | 6 - 7 | 814 | 100 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |