Assay Details
| Gene Symbol: NR4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712187.3 | XP_002712233.1 | 7 - 8 | 1948 | 61 | |
| XM_008258593.2 | XP_008256815.1 | 8 - 9 | 2012 | 61 | ||
| XM_008258594.2 | XP_008256816.1 | 7 - 8 | 1770 | 61 | ||
| XM_017342855.1 | XP_017198344.1 | 6 - 7 | 1828 | 61 | ||
| XM_017342856.1 | XP_017198345.1 | 7 - 8 | 1930 | 61 | ||
| XM_017342857.1 | XP_017198346.1 | 6 - 7 | 1648 | 61 | ||
| XM_017342858.1 | XP_017198347.1 | 7 - 8 | 1846 | 61 | ||
| XM_017342859.1 | XP_017198348.1 | 7 - 8 | 1772 | 61 | ||
More Information
Set Membership: |
|

NR4A2
Genomic Map