Assay Details
| Gene Symbol: SCARA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709303.3 | XP_002709349.2 | 5 - 6 | 1690 | 77 | |
| XM_017337935.1 | XP_017193424.1 | 4 - 5 | 1571 | 77 | ||
More Information
Set Membership: |
|

SCARA3
Genomic Map