Assay Details
| Gene Symbol: SERINC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344548.1 | XP_017200037.1 | 7 - 8 | 1113 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SERINC5| Gene Symbol: SERINC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344548.1 | XP_017200037.1 | 7 - 8 | 1113 | 61 | |
Set Membership: |
|