Assay Details
| Gene Symbol: ARFIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708762.3 | XP_002708808.1 | 6 - 7 | 863 | 85 | |
| XM_017344937.1 | XP_017200426.1 | 6 - 7 | 891 | 85 | ||
| XM_017344942.1 | XP_017200431.1 | 5 - 6 | 669 | 85 | ||
| XM_017344946.1 | XP_017200435.1 | 3 - 4 | 439 | 85 | ||
More Information
Set Membership: |
|

ARFIP2
Genomic Map