Assay Details
| Gene Symbol: TENT5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349678.1 | XP_017205167.1 | 2 - 3 | 176 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017349678.1 transcripts.
Set Membership: |
|

TENT5D
Genomic Map