Assay Details
| Gene Symbol: CDIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717993.3 | XP_002718039.1 | 4 - 5 | 532 | 76 | |
| XM_008269241.2 | XP_008267463.1 | 4 - 5 | 533 | 76 | ||
| XM_017348155.1 | XP_017203644.1 | 4 - 5 | 535 | 76 | ||
More Information
Set Membership: |
|

CDIN1
Genomic Map