Assay Details
| Gene Symbol: CDIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717993.3 | XP_002718039.1 | 5 - 6 | 612 | 90 | |
| XM_008269241.2 | XP_008267463.1 | 5 - 6 | 613 | 90 | ||
| XM_017348155.1 | XP_017203644.1 | 5 - 6 | 615 | 90 | ||
More Information
Set Membership: |
|

CDIN1
Genomic Map