Assay Details
| Gene Symbol: NMU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267849.2 | XP_008266071.1 | 2 - 3 | 505 | 71 | |
| XM_017347543.1 | XP_017203032.1 | 2 - 3 | 673 | 71 | ||
| XM_017347544.1 | XP_017203033.1 | 2 - 3 | 496 | 71 | ||
More Information
Set Membership: |
|

NMU
Genomic Map