Assay Details
| Gene Symbol: EHHADH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716469.3 | XP_002716515.1 | 6 - 7 | 972 | 58 | |
| XM_017346946.1 | XP_017202435.1 | 4 - 5 | 958 | 58 | ||
More Information
Set Membership: |
|

EHHADH
Genomic Map