Assay Details
| Gene Symbol: DNTTIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715851.3 | XP_002715897.1 | 6 - 7 | 2643 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNTTIP2| Gene Symbol: DNTTIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715851.3 | XP_002715897.1 | 6 - 7 | 2643 | 67 | |
Set Membership: |
|