Assay Details
Gene Symbol: TRIM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002721966.3 | XP_002722012.1 | 2110 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
