Assay Details
| Gene Symbol: LOC100354761 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709566.3 | XP_002709612.1 | 6 - 7 | 1014 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100354761| Gene Symbol: LOC100354761 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709566.3 | XP_002709612.1 | 6 - 7 | 1014 | 66 | |
Set Membership: |
|