Assay Details
| Gene Symbol: SCML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272398.2 | XP_008270620.1 | 1 - 2 | 466 | 69 | |
| XM_017350030.1 | XP_017205519.1 | 1 - 2 | 466 | 69 | ||
More Information
Set Membership: |
|

SCML1
Genomic Map