Assay Details
| Gene Symbol: ATG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261888.2 | XP_008260110.1 | 4 - 5 | 465 | 68 | |
| XM_008261889.2 | XP_008260111.1 | 5 - 6 | 581 | 68 | ||
| XM_008261891.2 | XP_008260113.1 | 4 - 5 | 490 | 68 | ||
| XM_017344554.1 | XP_017200043.1 | 6 - 7 | 574 | 68 | ||
| XM_017344555.1 | XP_017200044.1 | 6 - 7 | 575 | 68 | ||
| XM_017344556.1 | XP_017200045.1 | 6 - 7 | 574 | 68 | ||
| XM_017344557.1 | XP_017200046.1 | 6 - 7 | 575 | 68 | ||
| XM_017344558.1 | XP_017200047.1 | 3 - 4 | 370 | 68 | ||
| XM_017344559.1 | XP_017200048.1 | 4 - 5 | 459 | 68 | ||
| XM_017344560.1 | XP_017200049.1 | 2 - 3 | 262 | 68 | ||
More Information
Set Membership: |
|

ATG10
Genomic Map