Assay Details
| Gene Symbol: CDK5R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343226.1 | XP_017198715.1 | 1 | 336 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDK5R2| Gene Symbol: CDK5R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343226.1 | XP_017198715.1 | 1 | 336 | 115 | |
Set Membership: |
|