Assay Details
| Gene Symbol: OTUD6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720076.3 | XP_002720122.1 | 1 | 669 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OTUD6A| Gene Symbol: OTUD6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720076.3 | XP_002720122.1 | 1 | 669 | 81 | |
Set Membership: |
|