Assay Details
| Gene Symbol: BLM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008275317.2 | XP_008273539.1 | 994 | 81 | ||
| XM_008275320.2 | XP_008273542.1 | 994 | 81 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: BLM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008275317.2 | XP_008273539.1 | 994 | 81 | ||
| XM_008275320.2 | XP_008273542.1 | 994 | 81 | |||
Set Membership: |
|