Assay Details
| Gene Symbol: THG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710341.3 | XP_002710387.1 | 5 - 6 | 790 | 63 | |
| XM_017341224.1 | XP_017196713.1 | 5 - 6 | 578 | 63 | ||
More Information
Set Membership: |
|

THG1L
Genomic Map