Assay Details
| Gene Symbol: TMEM35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008273709.2 | XP_008271931.1 | 203 | 95 | ||
| XM_017350250.1 | XP_017205739.1 | 276 | 95 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM35B| Gene Symbol: TMEM35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008273709.2 | XP_008271931.1 | 203 | 95 | ||
| XM_017350250.1 | XP_017205739.1 | 276 | 95 | |||
Set Membership: |
|