Assay Details
| Gene Symbol: LOC100345461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248352.2 | XP_008246574.1 | 1170 | 63 | ||
| XM_017337618.1 | XP_017193107.1 | 1170 | 63 | |||
More Information
Set Membership: |
|

LOC100345461
Genomic Map