Assay Details
| Gene Symbol: GPRC5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257774.2 | XP_008255996.1 | 2 - 3 | 1221 | 66 | |
| XM_008257775.1 | XP_008255997.1 | 2 - 3 | 1165 | 66 | ||
| XM_008257777.2 | XP_008255999.1 | 2 - 3 | 1162 | 66 | ||
More Information
Set Membership: |
|

GPRC5B
Genomic Map