Assay Details
| Gene Symbol: PAGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711862.3 | XP_002711908.1 | 2 - 3 | 1258 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAGR1| Gene Symbol: PAGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711862.3 | XP_002711908.1 | 2 - 3 | 1258 | 71 | |
Set Membership: |
|