Assay Details
| Gene Symbol: IMPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710658.3 | XP_002710704.1 | 8 - 9 | 872 | 68 | |
| XM_017341415.1 | XP_017196904.1 | 8 - 9 | 871 | 68 | ||
| XM_017341416.1 | XP_017196905.1 | 7 - 8 | 864 | 68 | ||
More Information
Set Membership: |
|

IMPA1
Genomic Map