Assay Details
| Gene Symbol: PLPPR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715841.3 | XP_002715887.1 | 1 - 2 | 653 | 59 | |
| XM_017346057.1 | XP_017201546.1 | 1 - 2 | 654 | 59 | ||
More Information
Set Membership: |
|

PLPPR4
Genomic Map