Assay Details
| Gene Symbol: GOSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718900.3 | XP_002718946.1 | 1 - 2 | 133 | 69 | |
| XM_008270945.2 | XP_008269167.1 | 1 - 2 | 133 | 69 | ||
| XM_008270947.2 | XP_008269169.1 | 1 - 2 | 133 | 69 | ||
| XM_008270948.2 | XP_008269170.1 | 1 - 2 | 133 | 69 | ||
| XM_008270949.2 | XP_008269171.1 | 1 - 2 | 133 | 69 | ||
| XM_008270950.2 | XP_008269172.1 | 1 - 2 | 133 | 69 | ||
| XM_017349027.1 | XP_017204516.1 | 1 - 2 | 133 | 69 | ||
More Information
Set Membership: |
|

GOSR1
Genomic Map