Assay Details
| Gene Symbol: NARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708668.3 | XP_002708714.1 | 11 - 12 | 1421 | 85 | |
| XM_008263396.2 | XP_008261618.1 | 7 - 8 | 818 | 85 | ||
| XM_017344377.1 | XP_017199866.1 | 11 - 12 | 1422 | 85 | ||
| XM_017344379.1 | XP_017199868.1 | 9 - 10 | 1318 | 85 | ||
| XM_017344384.1 | XP_017199873.1 | 10 - 11 | 1280 | 85 | ||
| XM_017344386.1 | XP_017199875.1 | 7 - 8 | 675 | 85 | ||
| XM_017344389.1 | XP_017199878.1 | 7 - 8 | 639 | 85 | ||
More Information
Set Membership: |
|

NARS2
Genomic Map