Assay Details
| Gene Symbol: SEC22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716617.3 | XP_002716663.1 | 4 - 5 | 676 | 65 | |
| XM_008266833.2 | XP_008265055.1 | 4 - 5 | 676 | 65 | ||
| XM_017347052.1 | XP_017202541.1 | 4 - 5 | 676 | 65 | ||
More Information
Set Membership: |
|

SEC22A
Genomic Map