Assay Details
| Gene Symbol: POLL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270385.2 | XP_008268607.1 | 6 - 7 | 1167 | 53 | |
| XM_008270386.2 | XP_008268608.1 | 7 - 8 | 1342 | 53 | ||
| XM_017348506.1 | XP_017203995.1 | 8 - 9 | 1511 | 53 | ||
| XM_017348507.1 | XP_017203996.1 | 7 - 8 | 1346 | 53 | ||
| XM_017348508.1 | XP_017203997.1 | 4 - 5 | 707 | 53 | ||
More Information
Set Membership: |
|

POLL
Genomic Map