Assay Details
| Gene Symbol: NPFFR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348647.1 | XP_017204136.1 | 1 | 131 | 70 | |
| GenBank mRNA | JN665043.1 | - | 1 | 51 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPFFR1| Gene Symbol: NPFFR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348647.1 | XP_017204136.1 | 1 | 131 | 70 | |
| GenBank mRNA | JN665043.1 | - | 1 | 51 | 70 | |
Set Membership: |
|