Assay Details
| Gene Symbol: ERP27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712642.3 | XP_002712688.1 | 1 - 2 | 337 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERP27| Gene Symbol: ERP27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712642.3 | XP_002712688.1 | 1 - 2 | 337 | 73 | |
Set Membership: |
|