Assay Details
| Gene Symbol: LOC100351287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272625.1 | XP_008270847.1 | 1 | 856 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100351287| Gene Symbol: LOC100351287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272625.1 | XP_008270847.1 | 1 | 856 | 97 | |
Set Membership: |
|