Assay Details
| Gene Symbol: ARID5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017339929.1 | XP_017195418.1 | 3 - 4 | 275 | 55 | |
| XM_017339933.1 | XP_017195422.1 | 3 - 4 | 242 | 55 | ||
| XM_017339945.1 | XP_017195434.1 | 5 - 6 | 501 | 55 | ||
More Information
Set Membership: |
|

ARID5A
Genomic Map