Assay Details
| Gene Symbol: CAPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256803.2 | XP_008255025.1 | 7 - 8 | 775 | 77 | |
| XM_008256808.2 | XP_008255030.1 | 4 - 5 | 427 | 77 | ||
| XM_017341986.1 | XP_017197475.1 | 7 - 8 | 780 | 77 | ||
More Information
Set Membership: |
|

CAPS2
Genomic Map