Assay Details
| Gene Symbol: OXNAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716189.3 | XP_002716235.1 | 4 - 5 | 628 | 64 | |
| XM_008266044.2 | XP_008264266.1 | 4 - 5 | 678 | 64 | ||
More Information
Set Membership: |
|

OXNAD1
Genomic Map