Assay Details
| Gene Symbol: EEF1AKNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715894.3 | XP_002715940.1 | 4 - 5 | 2802 | 59 | |
| XM_008264068.2 | XP_008262290.1 | 5 - 6 | 2588 | 59 | ||
| XM_008264069.2 | XP_008262291.1 | 4 - 5 | 1295 | 59 | ||
| XM_008264070.2 | XP_008262292.1 | 4 - 5 | 1273 | 59 | ||
More Information
Set Membership: |
|

EEF1AKNMT
Genomic Map