Assay Details
| Gene Symbol: SUPT3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262998.2 | XP_008261220.1 | 11 - 12 | 1079 | 84 | |
| XM_008262999.2 | XP_008261221.1 | 10 - 11 | 992 | 84 | ||
More Information
Set Membership: |
|

SUPT3H
Genomic Map