Assay Details
Gene Symbol: CLDN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002722780.3 | XP_002722826.2 | 1029 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
