Assay Details
| Gene Symbol: NEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274196.2 | XP_008272418.1 | 245 | 66 | ||
| XM_017350495.1 | XP_017205984.1 | 251 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEMP1| Gene Symbol: NEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274196.2 | XP_008272418.1 | 245 | 66 | ||
| XM_017350495.1 | XP_017205984.1 | 251 | 66 | |||
Set Membership: |
|