Assay Details
| Gene Symbol: LOC100347361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017338514.1 | XP_017194003.1 | 688 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100347361| Gene Symbol: LOC100347361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017338514.1 | XP_017194003.1 | 688 | 88 | ||
Set Membership: |
|