Assay Details
| Gene Symbol: ST3GAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710551.3 | XP_002710597.1 | 7 - 8 | 1602 | 56 | |
| XM_017341545.1 | XP_017197034.1 | 6 - 7 | 1247 | 56 | ||
More Information
Set Membership: |
|

ST3GAL1
Genomic Map