Assay Details
| Gene Symbol: TUT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708251.3 | XP_002708297.3 | 22 - 23 | 4094 | 72 | |
| XM_008257414.2 | XP_008255636.2 | 22 - 23 | 4094 | 72 | ||
| XM_008257422.2 | XP_008255644.2 | 22 - 23 | 4093 | 72 | ||
| XM_008257427.2 | XP_008255649.2 | 22 - 23 | 4094 | 72 | ||
| XM_008257432.2 | XP_008255654.1 | 22 - 23 | 3998 | 72 | ||
More Information
Set Membership: |
|

TUT7
Genomic Map