Assay Details
| Gene Symbol: RGS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717695.3 | XP_002717741.1 | 4 - 5 | 610 | 107 | |
| XM_008268733.2 | XP_008266955.1 | 3 - 4 | 508 | 107 | ||
More Information
Set Membership: |
|

RGS18
Genomic Map