Assay Details
| Gene Symbol: BCL2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722959.3 | XP_002723005.2 | 615 | 113 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BCL2A1| Gene Symbol: BCL2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722959.3 | XP_002723005.2 | 615 | 113 | ||
Set Membership: |
|