Assay Details
| Gene Symbol: HNRNPH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718455.2 | XP_002718501.1 | 5 - 6 | 736 | 73 | |
| XM_008270053.1 | XP_008268275.1 | 5 - 6 | 755 | 73 | ||
| XM_008270054.2 | XP_008268276.1 | 5 - 6 | 713 | 73 | ||
| XM_008270055.1 | XP_008268277.1 | 5 - 6 | 710 | 73 | ||
| XM_008270056.2 | XP_008268278.1 | 5 - 6 | 713 | 73 | ||
| XM_017348741.1 | XP_017204230.1 | 5 - 6 | 758 | 73 | ||
| XM_017348742.1 | XP_017204231.1 | 3 - 4 | 430 | 73 | ||
| XM_017348743.1 | XP_017204232.1 | 3 - 4 | 401 | 73 | ||
More Information
Set Membership: |
|

HNRNPH3
Genomic Map