Assay Details
| Gene Symbol: GPKOW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719871.3 | XP_002719917.1 | 8 - 9 | 1197 | 66 | |
| XM_008272608.2 | XP_008270830.1 | 9 - 10 | 1142 | 66 | ||
| XM_017349756.1 | XP_017205245.1 | 8 - 9 | 1107 | 66 | ||
More Information
Set Membership: |
|

GPKOW
Genomic Map