Assay Details
| Gene Symbol: LOC100353768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257490.2 | XP_008255712.1 | 6 - 7 | 1286 | 64 | |
| XM_008257492.2 | XP_008255714.1 | 6 - 7 | 990 | 64 | ||
| XM_008257493.2 | XP_008255715.1 | 6 - 7 | 1297 | 64 | ||
| XM_017342308.1 | XP_017197797.1 | 6 - 7 | 1278 | 64 | ||
| XM_017342309.1 | XP_017197798.1 | 5 - 6 | 685 | 64 | ||
| XM_017342310.1 | XP_017197799.1 | 6 - 7 | 864 | 64 | ||
More Information
Set Membership: |
|

LOC100353768
Genomic Map