Assay Details
| Gene Symbol: LOC100353768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257490.2 | XP_008255712.1 | 9 - 10 | 1629 | 80 | |
| XM_008257492.2 | XP_008255714.1 | 9 - 10 | 1333 | 80 | ||
| XM_008257493.2 | XP_008255715.1 | 9 - 10 | 1640 | 80 | ||
| XM_017342308.1 | XP_017197797.1 | 9 - 10 | 1621 | 80 | ||
| XM_017342309.1 | XP_017197798.1 | 8 - 9 | 1028 | 80 | ||
| XM_017342310.1 | XP_017197799.1 | 9 - 10 | 1207 | 80 | ||
More Information
Set Membership: |
|

LOC100353768
Genomic Map